Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177806.3 | NP_808474.2 | 9 - 10 | 1458 | 69 | |
GenBank mRNA | AK154170.1 | - | 10 - 11 | 2082 | 69 | |
AK168462.1 | - | 9 - 10 | 1454 | 69 | ||
BC014689.1 | - | 9 - 10 | 1437 | 69 | ||
BC029153.1 | - | 4 - 5 | 430 | 69 | ||
BC030904.1 | - | 9 - 10 | 1400 | 69 | ||
BC033575.1 | - | 4 - 5 | 3525 | 69 | ||
BC076622.1 | - | 9 - 10 | 1144 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |