Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001178038.1 | NP_001171509.1 | 6 - 7 | 1285 | 98 | |
GenBank mRNA | BC051402.1 | - | 7 - 8 | 1372 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |