Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rc3h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024952.2 | NP_001020123.1 | 4 - 5 | 1029 | 78 | |
GenBank mRNA | AK139648.1 | - | 4 - 5 | 1001 | 78 | |
AK173335.1 | - | 4 - 5 | 975 | 78 | ||
AY948287.1 | - | 4 - 5 | 970 | 78 | ||
BC138663.1 | - | 3 - 4 | 753 | 8861684 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |