Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabgap1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013862.5 | NP_038890.3 | 16 - 17 | 2272 | 162 | |
GenBank mRNA | AK035796.1 | - | 16 - 17 | 2273 | 162 | |
BC145811.1 | - | 16 - 17 | 2117 | 40131114 | 162 | |
BC145813.1 | - | 16 - 17 | 2117 | 40131116 | 162 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |