Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081027.2 | NP_001074496.2 | 13 - 14 | 1289 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Ion Channel Mus musculus Probe spans exons |