Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110146.1 | NP_001103616.1 | 6 - 7 | 510 | 76 | |
NM_021400.3 | NP_067375.3 | 7 - 8 | 2904 | 76 | ||
GenBank mRNA | AK132597.1 | - | 1 - 2 | 650 | 76 | |
AK149469.1 | - | 6 - 7 | 511 | 76 | ||
BC130021.1 | - | 5 - 6 | 474 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |