Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177644.5 | NP_808312.3 | 6 - 7 | 1477 | 87 | |
GenBank mRNA | AK039584.1 | - | 6 - 7 | 1477 | 87 | |
AK086812.1 | - | 6 - 7 | 1103 | 87 | ||
BC120902.2 | - | 6 - 7 | 820 | 40086771 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |