Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081386.1 | NP_001074855.1 | 10 - 11 | 1809 | 142 | |
GenBank mRNA | BC157927.1 | - | 10 - 11 | 1800 | 9088100 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |