Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243223.1 | NP_001230152.1 | 14 - 15 | 2505 | 75 | |
GenBank mRNA | AK029630.1 | - | 5 - 6 | 612 | 75 | |
AK161463.1 | - | 7 - 8 | 925 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Mus musculus Probe spans exons |