Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100024871.mm.1 |
NM_001276767.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024871.mm.1 |
NM_144530.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024871.mm.1 |
TC0100003093.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100024867.mm.1 |
NM_001276767.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024867.mm.1 |
NM_144530.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024867.mm.1 |
TC0100003093.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100024869.mm.1 |
NM_001276767.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024869.mm.1 |
NM_144530.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024869.mm.1 |
TC0100003093.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100024870.mm.1 |
NM_001276767.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024870.mm.1 |
NM_144530.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024870.mm.1 |
TC0100003093.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100024866.mm.1 |
NM_001276767.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024866.mm.1 |
NM_144530.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024866.mm.1 |
TC0100003093.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100024865.mm.1 |
NM_001276767.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024865.mm.1 |
NM_144530.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024865.mm.1 |
TC0100003093.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100024872.mm.1 |
NM_001276767.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024872.mm.1 |
NM_144530.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024872.mm.1 |
TC0100003093.mm.1 |
MTA TC |