Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276767.1 | NP_001263696.1 | 7 - 8 | 913 | 81 | |
NM_144530.6 | NP_653113.4 | 6 - 7 | 859 | 81 | ||
GenBank mRNA | AK041148.1 | - | 6 - 7 | 833 | 81 | |
AK078105.1 | - | 6 - 7 | 822 | 81 | ||
AK122341.2 | - | 5 - 6 | 4394 | 81 | ||
AK134057.1 | - | 3 - 4 | 342 | 81 | ||
AK145740.1 | - | 6 - 7 | 832 | 81 | ||
AK167700.1 | - | 6 - 7 | 833 | 81 | ||
BC005786.1 | - | 5 - 6 | 676 | 2811511 | 81 | |
BC058552.1 | - | 5 - 6 | 918 | 81 | ||
BC066163.1 | - | 7 - 8 | 871 | 30092453 | 81 | |
BC066848.1 | - | 5 - 6 | 1670 | 30471872 | 81 | |
FR682665.1 | - | 7 - 8 | 832 | 81 |
Gene Symbol: Zbed6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | FR682664.1 | - | 6671 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |