Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf3ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028718.2 | NP_082994.1 | 4 - 5 | 590 | 72 | |
GenBank mRNA | AK014457.1 | - | 1 - 2 | 71 | 72 | |
AY613437.1 | - | 4 - 5 | 593 | 72 | ||
BC046538.1 | - | 4 - 5 | 593 | 72 | ||
BC117868.2 | - | 4 - 5 | 564 | 40095476 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |