Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dstyk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172516.4 | NP_766104.2 | 9 - 10 | 2384 | 85 | |
GenBank mRNA | AK050542.1 | - | 5 - 6 | 630 | 85 | |
AK083696.1 | - | 9 - 10 | 2282 | 85 | ||
AK132119.1 | - | 9 - 10 | 2259 | 85 | ||
AK145449.1 | - | 9 - 10 | 2229 | 85 | ||
AK172954.1 | - | 9 - 10 | 2230 | 85 | ||
AY208852.1 | - | 9 - 10 | 2231 | 85 | ||
AY429676.1 | - | 9 - 10 | 2270 | 85 | ||
BC019133.1 | - | 7 - 8 | 1019 | 85 | ||
BC089380.1 | - | 6 - 7 | 801 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |