Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nuak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195025.1 | NP_001181954.1 | 4 - 5 | 647 | 56 | |
NM_028778.4 | NP_083054.1 | 3 - 4 | 623 | 56 | ||
GenBank mRNA | AK004737.2 | - | 3 - 4 | 611 | 56 | |
AK033672.1 | - | 3 - 4 | 610 | 56 | ||
AK034082.1 | - | 4 - 5 | 639 | 56 | ||
BC046833.1 | - | 4 - 5 | 622 | 6816569 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |