Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053015.3 | NP_443748.2 | 12 - 13 | 1684 | 72 | |
GenBank mRNA | AB057759.1 | - | 11 - 12 | 1506 | 72 | |
AF384098.1 | - | 12 - 13 | 1681 | 72 | ||
AK019883.1 | - | 12 - 13 | 1655 | 72 | ||
AK148274.1 | - | 12 - 13 | 72 | |||
BC006900.1 | - | 12 - 13 | 1677 | 72 | ||
BC150804.1 | - | 12 - 13 | 1578 | 9087715 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |