Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136104.1 | NP_001129576.1 | 6 - 7 | 1309 | 102 | |
NM_009595.3 | NP_033725.2 | 6 - 7 | 1309 | 102 | ||
GenBank mRNA | BC137771.1 | - | 6 - 7 | 1109 | 8860790 | 102 |
BC144896.1 | - | 6 - 7 | 1109 | 102 | ||
BC144897.1 | - | 6 - 7 | 1109 | 9053445 | 102 | |
DQ084361.1 | - | 6 - 7 | 1047 | 102 | ||
U40827.1 | - | 2 - 3 | 124 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |