Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015811.2 | NP_056626.2 | 1 - 2 | 208 | 116 | |
GenBank mRNA | AF215667.1 | - | 1 - 2 | 139 | 116 | |
AK089293.1 | - | 1 - 2 | 164 | 116 | ||
AK138688.1 | - | 1 - 2 | 164 | 116 | ||
AK154394.1 | - | 164 | 116 | |||
AK170366.1 | - | 1 - 2 | 160 | 116 | ||
AK170957.1 | - | 1 - 2 | 164 | 116 | ||
BC028634.1 | - | 1 - 2 | 139 | 1328269 | 116 | |
BU695313.1 | - | 1 - 2 | 230 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |