Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113188.1 | NP_001106659.1 | 7 - 8 | 664 | 97 | |
NM_001113189.1 | NP_001106660.1 | 7 - 8 | 664 | 97 | ||
NM_008053.2 | NP_032079.1 | 7 - 8 | 664 | 97 | ||
GenBank mRNA | AF124385.1 | - | 2 - 3 | 165 | 97 | |
BC019139.1 | - | 7 - 8 | 645 | 5041635 | 97 | |
X90875.1 | - | 7 - 8 | 643 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |