Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gigyf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110212.2 | NP_001103682.1 | 11 - 12 | 1281 | 88 | |
NM_146112.4 | NP_666224.3 | 11 - 12 | 1299 | 88 | ||
GenBank mRNA | AK079246.1 | - | 11 - 12 | 1251 | 88 | |
AK122337.1 | - | 2 - 3 | 370 | 88 | ||
AK144058.1 | - | 11 - 12 | 1261 | 88 | ||
AK157733.1 | - | 2 - 3 | 2373 | 88 | ||
AK160878.1 | - | 9 - 10 | 1093 | 88 | ||
AY176043.1 | - | 11 - 12 | 1245 | 88 | ||
BC076610.1 | - | 11 - 12 | 1244 | 88 | ||
BC089036.1 | - | 11 - 12 | 1255 | 30695300 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |