Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009118.2 | NP_033144.1 | 4 - 5 | 551 | 77 | |
GenBank mRNA | AK044214.1 | - | 3 - 4 | 232 | 77 | |
AK053278.1 | - | 4 - 5 | 310 | 77 | ||
AK143098.1 | - | 4 - 5 | 310 | 77 | ||
AK149299.1 | - | 4 - 5 | 560 | 77 | ||
AY651760.1 | - | 4 - 5 | 339 | 77 | ||
BC016498.1 | - | 4 - 5 | 344 | 4527551 | 77 | |
M24086.1 | - | 4 - 5 | 338 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |