Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efhc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028916.4 | NP_083192.2 | 10 - 11 | 1698 | 138 | |
GenBank mRNA | AK016714.1 | - | 10 - 11 | 1657 | 138 | |
AK029501.1 | - | 10 - 11 | 1664 | 138 | ||
BC120659.1 | - | 10 - 11 | 1633 | 40129582 | 138 | |
BC125625.1 | - | 10 - 11 | 1633 | 40130140 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |