Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efhc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028916.4 | NP_083192.2 | 1 - 2 | 128 | 65 | |
GenBank mRNA | AK016714.1 | - | 1 - 2 | 87 | 65 | |
AK029501.1 | - | 1 - 2 | 94 | 65 | ||
BC120659.1 | - | 1 - 2 | 63 | 40129582 | 65 | |
BC125625.1 | - | 1 - 2 | 63 | 40130140 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |