Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btnl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172793.2 | NP_766381.1 | 1 - 2 | 104 | 90 | |
GenBank mRNA | AK080918.1 | - | 1 - 2 | 87 | 90 | |
AK084528.1 | - | 1 - 2 | 105 | 90 | ||
AY177687.1 | - | 1 - 2 | 71 | 90 | ||
BC025540.1 | - | 1 - 2 | 82 | 90 | ||
BC150808.1 | - | 1 - 2 | 61 | 9087719 | 90 | |
BC151155.1 | - | 1 - 2 | 61 | 9088043 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |