Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfhx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030708.2 | NP_109633.2 | 8 - 9 | 4178 | 98 | |
GenBank mRNA | AB024499.1 | - | 7 - 8 | 3802 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |