Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008862.3 | NP_032888.1 | 1 - 2 | 148 | 75 | |
GenBank mRNA | AK139347.1 | - | 1 - 2 | 80 | 75 | |
BC048244.1 | - | 1 - 2 | 82 | 6308831 | 75 | |
BY253073.1 | - | 1 - 2 | 149 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |