Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027375.2 | NP_081651.2 | 16 - 17 | 3947 | 120 | |
GenBank mRNA | AB093232.1 | - | 16 - 17 | 3950 | 120 | |
BC023811.1 | - | 11 - 12 | 1334 | 120 | ||
BC034077.1 | - | 5 - 6 | 585 | 120 | ||
BC065697.1 | - | 4 - 5 | 342 | 120 | ||
BC139007.1 | - | 16 - 17 | 3943 | 8862029 | 120 | |
BC145495.1 | - | 16 - 17 | 4016 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |