Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252494.1 | NP_001239423.1 | 25 - 26 | 3961 | 84 | |
NM_001252496.1 | NP_001239425.1 | 26 - 27 | 3985 | 84 | ||
NM_175258.4 | NP_780467.2 | 24 - 25 | 3946 | 84 | ||
GenBank mRNA | AK143331.1 | - | 12 - 13 | 2333 | 84 | |
AK220459.1 | - | 14 - 15 | 2373 | 84 | ||
BC019702.1 | - | 3 - 4 | 523 | 84 | ||
BC025553.1 | - | 7 - 8 | 1440 | 84 | ||
BC141182.1 | - | 25 - 26 | 3860 | 9055794 | 84 | |
BC145503.1 | - | 26 - 27 | 3885 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |