Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173753.4 | NP_776114.2 | 6 - 7 | 740 | 82 | |
GenBank mRNA | AK087849.1 | - | 6 - 7 | 1394 | 82 | |
AK122574.1 | - | 6 - 7 | 688 | 82 | ||
BC079892.1 | - | 6 - 7 | 730 | 30543296 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |