Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173753.4 | NP_776114.2 | 7 - 8 | 822 | 69 | |
GenBank mRNA | AK087849.1 | - | 7 - 8 | 1476 | 69 | |
AK122574.1 | - | 7 - 8 | 770 | 69 | ||
BC079892.1 | - | 7 - 8 | 812 | 30543296 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |