Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290805.1 | NP_001277734.1 | 3 - 4 | 578 | 86 | |
NM_001290806.1 | NP_001277735.1 | 3 - 4 | 578 | 86 | ||
NM_008443.4 | NP_032469.2 | 3 - 4 | 578 | 86 | ||
GenBank mRNA | AK147078.1 | - | 3 - 4 | 528 | 86 | |
AK161223.1 | - | 3 - 4 | 569 | 86 | ||
AK161920.1 | - | 3 - 4 | 552 | 86 | ||
AK169425.1 | - | 3 - 4 | 552 | 86 | ||
BC023936.1 | - | 3 - 4 | 528 | 86 | ||
BC044720.1 | - | 3 - 4 | 539 | 86 | ||
BC052707.1 | - | 3 - 4 | 544 | 5702415 | 86 | |
D12645.1 | - | 3 - 4 | 554 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |