Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmnd5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025346.1 | NP_079622.1 | 8 - 9 | 1317 | 102 | |
GenBank mRNA | AK005344.1 | - | 5 - 6 | 696 | 102 | |
AK075599.1 | - | 8 - 9 | 1317 | 102 | ||
BC016075.1 | - | 8 - 9 | 1171 | 4485143 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |