Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048061.1 | NP_001041526.1 | 7 - 8 | 1165 | 83 | |
GenBank mRNA | AK165609.1 | - | 8 - 9 | 1112 | 83 | |
AY137376.1 | - | 3 - 4 | 282 | 83 | ||
BQ445737.1 | - | 4 - 5 | 525 | 83 | ||
DQ397821.1 | - | 6 - 7 | 927 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Probe spans exons |