Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166654.1 | NP_001160126.1 | 12 - 13 | 2037 | 155 | |
GenBank mRNA | AK046394.1 | - | 12 - 13 | 2038 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Mus musculus Probe spans exons |