Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163671.1 | NP_001157143.1 | 9 - 10 | 1273 | 65 | |
NM_001163672.1 | NP_001157144.1 | 9 - 10 | 1273 | 65 | ||
NM_016961.3 | NP_058657.1 | 9 - 10 | 1273 | 65 | ||
NM_207692.2 | NP_997575.2 | 9 - 10 | 1273 | 65 | ||
GenBank mRNA | AB005664.1 | - | 8 - 9 | 1047 | 65 | |
AF052466.1 | - | 8 - 9 | 1009 | 65 | ||
AF052467.1 | - | 8 - 9 | 1009 | 65 | ||
AF052468.1 | - | 8 - 9 | 1009 | 65 | ||
AF052469.1 | - | 8 - 9 | 1009 | 65 | ||
AK031959.1 | - | 9 - 10 | 1272 | 65 | ||
AK031974.1 | - | 7 - 8 | 827 | 65 | ||
AK145083.1 | - | 9 - 10 | 1248 | 65 | ||
BC028341.1 | - | 9 - 10 | 1255 | 4192796 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |