Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290709.1 | NP_001277638.1 | 6 - 7 | 630 | 101 | |
NM_001290710.1 | NP_001277639.1 | 6 - 7 | 630 | 101 | ||
NM_001290711.1 | NP_001277640.1 | 6 - 7 | 749 | 101 | ||
NM_007897.3 | NP_031923.1 | 6 - 7 | 630 | 101 | ||
GenBank mRNA | AK031239.1 | - | 6 - 7 | 625 | 101 | |
AK035781.1 | - | 6 - 7 | 513 | 101 | ||
AK137389.1 | - | 6 - 7 | 671 | 101 | ||
AK139021.1 | - | 6 - 7 | 628 | 101 | ||
AK141175.1 | - | 6 - 7 | 630 | 101 | ||
BC139362.1 | - | 6 - 7 | 591 | 8862384 | 101 | |
BC139364.1 | - | 6 - 7 | 591 | 8862386 | 101 | |
L12147.1 | - | 6 - 7 | 596 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |