Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290711.1 | NP_001277640.1 | 3 - 4 | 552 | 89 | |
GenBank mRNA | AK137389.1 | - | 3 - 4 | 474 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |