Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thg1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080969.3 | NP_001074438.1 | 4 - 5 | 661 | 109 | |
NM_001290737.1 | NP_001277666.1 | 5 - 6 | 604 | 109 | ||
GenBank mRNA | AK010876.1 | - | 4 - 5 | 653 | 109 | |
AK015467.1 | - | 4 - 5 | 455 | 109 | ||
AK016076.1 | - | 5 - 6 | 605 | 109 | ||
AK017532.1 | - | 4 - 5 | 644 | 109 | ||
BC115560.1 | - | 4 - 5 | 649 | 40101901 | 109 | |
BC115561.1 | - | 4 - 5 | 649 | 40101902 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |