Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045520.3 | NP_001038985.2 | 3 - 4 | 433 | 90 | |
GenBank mRNA | AB093212.1 | - | 2 - 3 | 1520 | 90 | |
AK143544.1 | - | 3 - 4 | 434 | 90 | ||
AK147875.1 | - | 3 - 4 | 418 | 90 | ||
AK168991.1 | - | 3 - 4 | 399 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |