Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctse | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007799.3 | NP_031825.2 | 1 - 2 | 249 | 111 | |
GenBank mRNA | AK145875.1 | - | 1 - 2 | 106 | 111 | |
BC005432.1 | - | 1 - 2 | 249 | 3601519 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus 1700 Probe spans exons |