Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Daam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286452.1 | NP_001273381.1 | 23 - 24 | 3162 | 88 | |
NM_026102.3 | NP_080378.2 | 24 - 25 | 3157 | 88 | ||
NM_172464.3 | NP_766052.2 | 24 - 25 | 3022 | 88 | ||
GenBank mRNA | AK129185.1 | - | 24 - 25 | 3000 | 88 | |
AK147480.1 | - | 23 - 24 | 2976 | 88 | ||
AY426535.1 | - | 24 - 25 | 3067 | 88 | ||
BC025638.1 | - | 22 - 23 | 2892 | 88 | ||
BC032287.1 | - | 10 - 11 | 1066 | 88 | ||
BC038403.1 | - | 23 - 24 | 2936 | 88 | ||
BC047431.1 | - | 24 - 25 | 2944 | 88 | ||
BC048856.1 | - | 4 - 5 | 485 | 88 | ||
BC076585.1 | - | 24 - 25 | 3012 | 30622306 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |