Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC119618.1 | - | 10 - 11 | 890 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |