Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019393.2 | NP_062266.1 | 9 - 10 | 1051 | 84 | |
GenBank mRNA | AF152841.1 | - | 9 - 10 | 1024 | 84 | |
AF152842.1 | - | 9 - 10 | 1073 | 84 | ||
AK010253.1 | - | 9 - 10 | 1025 | 84 | ||
AK011636.1 | - | 9 - 10 | 1002 | 84 | ||
AK076096.1 | - | 9 - 10 | 1031 | 84 | ||
BC005622.1 | - | 9 - 10 | 1030 | 3711930 | 84 | |
BC052156.1 | - | 9 - 10 | 1023 | 6394529 | 84 | |
BC158075.1 | - | 9 - 10 | 1002 | 9007372 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |