Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027810.3 | NP_082086.2 | 13 - 14 | 1474 | 74 | |
GenBank mRNA | AF521645.1 | - | 13 - 14 | 1473 | 74 | |
AK050291.1 | - | 13 - 14 | 1472 | 74 | ||
AK132661.1 | - | 13 - 14 | 1402 | 74 | ||
BC031505.1 | - | 13 - 14 | 1424 | 3494587 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |