Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164056.1 | NP_001157528.1 | 3 - 4 | 414 | 96 | |
NM_008875.4 | NP_032901.2 | 2 - 3 | 359 | 96 | ||
GenBank mRNA | BC068144.1 | - | 2 - 3 | 216 | 30532470 | 96 |
CF727963.1 | - | 2 - 3 | 359 | 96 | ||
U87868.1 | - | 3 - 4 | 397 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |