Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008875.4 | NP_032901.2 | 1 - 2 | 169 | 84 | |
GenBank mRNA | CF727963.1 | - | 1 - 2 | 169 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |