Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013531.4 | NP_038559.2 | 8 - 9 | 830 | 64 | |
GenBank mRNA | AF277893.1 | - | 7 - 8 | 694 | 64 | |
AK036816.1 | - | 8 - 9 | 832 | 64 | ||
AK098139.1 | - | 8 - 9 | 763 | 64 | ||
AK152227.1 | - | 8 - 9 | 821 | 64 | ||
AK155309.1 | - | 8 - 9 | 889 | 64 | ||
AK158965.1 | - | 9 - 10 | 1060 | 64 | ||
AK167416.1 | - | 8 - 9 | 863 | 64 | ||
AK168302.1 | - | 8 - 9 | 786 | 64 | ||
AK169752.1 | - | 7 - 8 | 907 | 64 | ||
AK172147.1 | - | 9 - 10 | 963 | 64 | ||
M63658.1 | - | 7 - 8 | 734 | 64 | ||
M87286.1 | - | 7 - 8 | 734 | 64 | ||
S86124.1 | - | 7 - 8 | 734 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |