Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024200.4 | NP_077162.2 | 11 - 12 | 1447 | 154 | |
GenBank mRNA | AK018181.1 | - | 11 - 12 | 1444 | 154 | |
AK081358.1 | - | 4 - 5 | 433 | 154 | ||
AK141611.1 | - | 11 - 12 | 1448 | 154 | ||
AK220169.1 | - | 11 - 12 | 1435 | 154 | ||
AY174062.1 | - | 10 - 11 | 1576 | 154 | ||
BC008137.1 | - | 7 - 8 | 714 | 154 | ||
BC047050.1 | - | 2 - 3 | 182 | 154 | ||
BC056641.1 | - | 11 - 12 | 1406 | 5097345 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |