Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024200.4 | NP_077162.2 | 13 - 14 | 1651 | 157 | |
GenBank mRNA | AK018181.1 | - | 13 - 14 | 1648 | 157 | |
AK081358.1 | - | 6 - 7 | 637 | 157 | ||
AK141611.1 | - | 13 - 14 | 1652 | 157 | ||
AK220169.1 | - | 13 - 14 | 1639 | 157 | ||
AY174062.1 | - | 12 - 13 | 1780 | 157 | ||
BC008137.1 | - | 9 - 10 | 918 | 157 | ||
BC047050.1 | - | 4 - 5 | 386 | 157 | ||
BC056641.1 | - | 13 - 14 | 1610 | 5097345 | 157 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |