Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027016.2 | NP_081292.1 | 1 - 2 | 511 | 67 | |
GenBank mRNA | AK013922.1 | - | 1 - 2 | 326 | 67 | |
AK088093.1 | - | 1 - 2 | 60 | 67 | ||
AK090078.1 | - | 1 - 2 | 61 | 67 | ||
BC017162.1 | - | 1 - 2 | 512 | 67 | ||
BC048532.1 | - | 1 - 2 | 61 | 67 | ||
BC053504.1 | - | 1 - 2 | 48 | 67 | ||
BC067202.1 | - | 1 - 2 | 46 | 6332701 | 67 | |
BC071243.1 | - | 1 - 2 | 61 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |