Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028140.4 | NP_082416.2 | 8 - 9 | 1006 | 97 | |
GenBank mRNA | AK013144.1 | - | 8 - 9 | 898 | 97 | |
AK032080.1 | - | 8 - 9 | 1027 | 97 | ||
AK035291.1 | - | 8 - 9 | 970 | 97 | ||
AK077957.1 | - | 8 - 9 | 1007 | 97 | ||
BC010483.1 | - | 8 - 9 | 855 | 97 | ||
BC113183.1 | - | 8 - 9 | 937 | 40061891 | 97 | |
BC113790.1 | - | 8 - 9 | 937 | 40061889 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |